Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Frizzled-10 Rabbit Polyclonal Antibody [clone: L164]
  BWRLBS2601
Anti-Frizzled-10 Rabbit Polyclonal Antibody [clone: L164]
  BWRLBS2601

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Frizzled-10
  • Clonality:
    Polyclonal
  • Clone:
    L164
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 132-180 of Human Frizzled-10.

Frizzled-10 (L164) pAb detects endogenous levels of Frizzled-10 protein.

Type: Primary
Antigen: Frizzled-10
Clonality: Polyclonal
Clone: L164
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse