Anti-IL-9R Rabbit Polyclonal Antibody [clone: L514]
Catalog # BWRLBS1669
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IL-9R
- Clonality:Polyclonal
- Clone:L514
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 480-530 of Human IL-9R.
IL-9R (L514) pAb detects endogenous levels of IL-9R protein.
Type: Primary
Antigen: IL-9R
Clonality: Polyclonal
Clone: L514
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human