Anti-Sox8/9/17/18 Rabbit Polyclonal Antibody [clone: K148]
Catalog # BWRLBS2481
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Sox8/9/17/18
- Clonality:Polyclonal
- Clone:K148
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 120-170 of Human Sox8.
Sox-8/9/17/18 (K148) pAb detects endogenous levels of Sox-8/9/17/18 protein.
Type: Primary
Antigen: Sox8/9/17/18
Clonality: Polyclonal
Clone: K148
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse