Anti-Sam 68 Rabbit Polyclonal Antibody [clone: Y145]
Catalog # BWRLBS2952
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Sam 68
- Clonality:Polyclonal
- Clone:Y145
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 120-170 of Human Sam 68.
Sam 68 (Y145) pAb detects endogenous levels of Sam 68 protein.
Type: Primary
Antigen: Sam 68
Clonality: Polyclonal
Clone: Y145
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat