Anti-c-Kit Rabbit Polyclonal Antibody [clone: E930]
BWRLBS2433
- Antibody type:Primary
- Antigen symbol:c-Kit
- Clonality:Polyclonal
- Clone:E930
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 901-950 of Human c-Kit.
c-Kit (E930) pAb detects endogenous levels of c-Kit protein.
Type: Primary
Antigen: c-Kit
Clonality: Polyclonal
Clone: E930
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse