Anti-Connexin 37 Rabbit Polyclonal Antibody [clone: L116]
Catalog # BWRLBS3866
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Connexin 37
- Clonality:Polyclonal
- Clone:L116
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 100-150 of Human Connexin 37.
Connexin 37 (L116) pAb detects endogenous levels of Connexin 37 protein.
Type: Primary
Antigen: Connexin 37
Clonality: Polyclonal
Clone: L116
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat