Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IFI-16 Rabbit Polyclonal Antibody [clone: N772]
Anti-IFI-16 Rabbit Polyclonal Antibody [clone: N772]
  BWRLBS3865
Anti-IFI-16 Rabbit Polyclonal Antibody [clone: N772]
  BWRLBS3865

 

  • Antibody type:
    Primary
  • Antigen symbol:
    IFI-16
  • Clonality:
    Polyclonal
  • Clone:
    N772
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 740-795 of Human IFI-16.

IFI-16 (N772) pAb detects endogenous levels of IFI-16 protein.

Type: Primary
Antigen: IFI-16
Clonality: Polyclonal
Clone: N772
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human