Anti-CLIC3 Rabbit Polyclonal Antibody [clone: F56]
Catalog # BWRLBS3832
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CLIC3
- Clonality:Polyclonal
- Clone:F56
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 31-80 of Human CLIC3.
CLIC3 (F56) pAb detects endogenous levels of CLIC3 protein.
Type: Primary
Antigen: CLIC3
Clonality: Polyclonal
Clone: F56
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat