Anti-Trypsin-3 Rabbit Polyclonal Antibody [clone: M166]
BWRLBS3707
- Antibody type:Primary
- Antigen symbol:Trypsin-3
- Clonality:Polyclonal
- Clone:M166
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 150-200 of Human Trypsin-3.
Trypsin-3 (M166) pAb detects endogenous levels of Trypsin-3 protein.
Type: Primary
Antigen: Trypsin-3
Clonality: Polyclonal
Clone: M166
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human