Anti-Cadherin-18 Rabbit Polyclonal Antibody [clone: Q129]
Catalog # BWRLBS2222
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cadherin-18
- Clonality:Polyclonal
- Clone:Q129
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 96-140 of Human Cadherin-18.
Cadherin-18 (Q129) pAb detects endogenous levels of Cadherin-18 protein.
Type: Primary
Antigen: Cadherin-18
Clonality: Polyclonal
Clone: Q129
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat