Anti-MMP-12 Rabbit Polyclonal Antibody [clone: D411]
Catalog # BWRLBS3699
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MMP-12
- Clonality:Polyclonal
- Clone:D411
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 400-450 of Human MMP-12.
MMP-12 (D411) pAb detects endogenous levels of MMP-12 protein.
Type: Primary
Antigen: MMP-12
Clonality: Polyclonal
Clone: D411
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human