Anti-IL-32 Rabbit Polyclonal Antibody [clone: K226]
Catalog # BWRLBS3676
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IL-32
- Clonality:Polyclonal
- Clone:K226
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 200-2450 of Human IL-32.
IL-32 (K226) pAb detects endogenous levels of IL-32 protein.
Type: Primary
Antigen: IL-32
Clonality: Polyclonal
Clone: K226
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human