Anti-GluR2 Rabbit Polyclonal Antibody [clone: Y873]
Catalog # BWRLBS3658
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GluR2
- Clonality:Polyclonal
- Clone:Y873
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids C-terminus of Human GluR2.
GluR2 (Y873) pAb detects endogenous levels of GluR2 protein.
Type: Primary
Antigen: GluR2
Clonality: Polyclonal
Clone: Y873
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat