Anti-14-3-3 epsilon Rabbit Polyclonal Antibody [clone: D238]
Catalog # BWRLBS2177
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 epsilon
- Clonality:Polyclonal
- Clone:D238
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 200-250 of Human 14-3-3 epsilon.
14-3-3 ? (D238) pAb detects endogenous levels of 14-3-3 ? protein.
Type: Primary
Antigen: 14-3-3 epsilon
Clonality: Polyclonal
Clone: D238
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat