Anti-DARPP-32 Rabbit Polyclonal Antibody [clone: P69]
BWRLBS1394
- Antibody type:Primary
- Antigen symbol:DARPP-32
- Clonality:Polyclonal
- Clone:P69
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 35-82 of Human DARPP-32.
DARPP-32 (P69) pAb detects endogenous levels of DARPP-32 protein.
Type: Primary
Antigen: DARPP-32
Clonality: Polyclonal
Clone: P69
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat