Anti-WBSCR11 Rabbit Polyclonal Antibody [clone: K94]
Catalog # BWRLBS2161
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:WBSCR11
- Clonality:Polyclonal
- Clone:K94
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 63-112 of Human WBSCR11.
WBSCR11 (K94) pAb detects endogenous levels of WBSCR11 protein.
Type: Primary
Antigen: WBSCR11
Clonality: Polyclonal
Clone: K94
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat