Anti-p-c-Jun Rabbit Polyclonal Antibody [clone: S63]
Catalog # BWRLBS4045
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-c-Jun
- Clonality:Polyclonal
- Clone:S63
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human c-Jun around the phosphorylation site of Serine 63.
p-c-Jun (S63) pAb detects endogenous levels of c-Jun protein only when phosphorylated at Ser63.
Type: Primary
Antigen: p-c-Jun
Clonality: Polyclonal
Clone: S63
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat