Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-p-ATF2 Rabbit Polyclonal Antibody [clone: S62]
  BWRLBS4016
Anti-p-ATF2 Rabbit Polyclonal Antibody [clone: S62]
  BWRLBS4016

 

  • Antibody type:
    Primary
  • Antigen symbol:
    p-ATF2
  • Clonality:
    Polyclonal
  • Clone:
    S62
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic phosphopeptide derived from human ATF2 around the phosphorylation site of Serine 62.

p-ATF2 (S62) pAb detects endogenous levels of ATF2 protein only when phosphorylated at Ser62.

Type: Primary
Antigen: p-ATF2
Clonality: Polyclonal
Clone: S62
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat