Anti-SRSF7 Rabbit Polyclonal Antibody [clone: N103]
Catalog # BWRLBS3954
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:SRSF7
- Clonality:Polyclonal
- Clone:N103
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 71-122 of Human SRSF7.
SRSF7 (N103) pAb detects endogenous levels of SRSF7 protein.
Type: Primary
Antigen: SRSF7
Clonality: Polyclonal
Clone: N103
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse