Anti-Connexin 32 Rabbit Polyclonal Antibody [clone: L115]
Catalog # BWRLBS3527
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Connexin 32
- Clonality:Polyclonal
- Clone:L115
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 81-130 of Human Connexin 32.
Connexin-32 (L115) pAb detects endogenous levels of Connexin-32 protein.
Type: Primary
Antigen: Connexin 32
Clonality: Polyclonal
Clone: L115
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat