Anti-TNF-beta Rabbit Polyclonal Antibody [clone: H66]
Catalog # BWRLBS3491
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:TNF- beta
- Clonality:Polyclonal
- Clone:H66
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 31-80 of Human TNF-beta.
TNF-? (H66) pAb detects endogenous levels of TNF-? protein.
Type: Primary
Antigen: TNF-beta
Clonality: Polyclonal
Clone: H66
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat