Anti-IL-20 Rabbit Polyclonal Antibody [clone: D61]
Catalog # BWRLBS3464
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IL-20
- Clonality:Polyclonal
- Clone:D61
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 30-78 of Human IL-20.
IL-20 (D61) pAb detects endogenous levels of IL-20 protein.
Type: Primary
Antigen: IL-20
Clonality: Polyclonal
Clone: D61
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse