Order Entry
Czech Republic
ContactUsLinkComponent
Hypoxanthine-Aminopterin-Thymidin (HAT) media supplement, Hybri-Max™
Hypoxanthine-Aminopterin-Thymidin (HAT) media supplement, Hybri-Max™
Catalog # SIALH0262-10VL
Supplier:  Merck
CAS Number:  
Hypoxanthine-Aminopterin-Thymidin (HAT) media supplement, Hybri-Max™
Catalog # SIALH0262-10VL
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 Vial
  • Description:
    HAT media supplement
  • Storage temperature:
    −20 °C

Specifications

About this item

Hypoxanthine-Aminopterin-Thymidine (HAT) medium is used for the fusion of myeloma cells with spleen cells and the selection of hybridomas (cell hybrids) in culture.

  • Lyophilised powder
  • Gamma-irradiated

Supplements are available in vials with 10 ml sterile culture medium, each one sufficient to supplement 500 ml of medium.