ContactUsLinkComponent
Weighing pans, aluminium
Weighing pans, aluminium
Catalog # KOEH2-1607
undefined
Weighing pans, aluminium
Catalog # KOEH2-1607
Specifications
  • Capacity:
    160 ml
  • Colour:
    Silver
  • Description:
    Rectangular aluminium shells with edge and conical tapering
  • W×D×H:
    64×100×38 mm
  • Pk:
    100
Specifications
About this item
Made of Aluminium, bright.
  • Good temperature resistance
  • Good chemical resistance
  • Lightweight
  • Stackable