Orders LinkContactUsLinkComponent
Safety spectacles, i-works
Safety spectacles, i-works
Supplier:   UVEX
Safety spectacles, i-works
Supplier:  UVEX
Certificates
Safety spectacles, i-works
Safety spectacles, i-works
4 Options Available Below
Product Details & Documents
The 100% metal free uvex i-works encapsulates the best of uvex eyewear: the sporty design, panoramic lens and low weight make it extremely comfortable and together with the unrivalled permanent uvex lens coating technology (uvex supravision excellence), provides the highest level of optical clarity remaining fog free even after repeated cleaning. High-tech soft grip side arm materials further enhance comfort levels delivering a stable, non-slip fit.
  • Sporty design and panoramic lens for great lateral vision
  • High performance anti-fog coating on the inside, scratch resistant on the outside
  • Comfortable stable, non-slip soft grip side arms
  • Low profile side arms with practical hang cord attachment option
  • 100% UV 400
Suitable for a range of applications requiring low impact protection including those working in construction, manufacturing, automotive, healthcare, utilities, pharmaceutical, food and beverages, warehouse and distribution.
Certifications: EN 166 (EN 172)
Documents
Product Family Options
Product Information
  • Type
    Colour
    Lenses
    Availability
    Price
  • Anti-UV (2C-1,2), anti-scratch, anti-fog
    Grey/blue
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 259,00 Kč
    259,00 Kč
    1 * 1 KS
       259,00 Kč
  • Anti-UV (2C-1,2), anti-scratch, anti-fog
    Grey/green
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 259,00 Kč
    259,00 Kč
    1 * 1 KS
       259,00 Kč
  • Anti-UV (2C-1,2), anti-scratch, anti-fog
    Black/yellow
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 307,00 Kč
    307,00 Kč
    1 * 1 KS
       307,00 Kč
  • Anti-UV (5-2,5), anti-scratch, anti-fog
    Anthracite
    Grey PC
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 285,00 Kč
    285,00 Kč
    1 * 1 KS
       285,00 Kč