ContactUsLinkComponent
VWR® VisiCam® TV Analog Camera
  630-1120
undefined
VWR® VisiCam® TV Analog Camera
  630-1120
  • Resolution:
    470 TVL
  • Sensor type:
    CCD
  • Sensor size:
    1/3"
  • Description:
    Analogic camera, EU-plug
  • Sensor:
    Colour CCD
  • Pk:
    1
Analog camera that can be connected to video projectors, external monitors or a computer via the optional USB set.
  • ¹/₃" CCD sensor (798×548 pixels)
  • White balance: automatic/manual
  • Resolution: 470 TV lines
 : Storage and analysis of images via the optional USB converter kit, which needs to be ordered separately.
 : Supplied with an eyepiece adapter, a CS-C mount adapter, a calibration slideand CCTV lens.