Order Entry
Czech Republic
ContactUsLinkComponent
Triethoxy-3-(2-imidazolin-1-yl)propylsilane ≥97% (by total nitrogen and titration analysis), liquid, Sigma-Aldrich®
Irritant
Triethoxy-3-(2-imidazolin-1-yl)propylsilane ≥97% (by total nitrogen and titration analysis), liquid, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Triethoxy-3-(2-imidazolin-1-yl)propylsilane ≥97% (by total nitrogen and titration analysis), liquid, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Appearance:
    Colorless to very dark yellow and colorless to very dark green-yellow
  • Form:
    Liquid
  • Titration (NT) HClO₄ 0,1 M:
    97,0 - 103,0%
  • Refractive index N20/D:
    1,451 - 1,453
  • Proton NMR spectrum:
    Conforms to structure

Specification Test Results

Frequently Bought Together