ContactUsLinkComponent
Serological pipettes, Corning®
Serological pipettes, Corning®
Catalog # CORN7374
Supplier:  Corning
undefined
Serological pipettes, Corning®
Catalog # CORN7374
Supplier:  Corning
Specifications
  • Packed:
    50/bag
  • Sterile:
    +
  • Capacity:
    25 ml
  • Seq:
    05
  • Pk:
    200
Specifications
About this item
Sterile serological pipettes made of PS.
Packaging: Triple bagged.
Caution: These pipettes are "Made To Order" products. The minimum order quantity is 40 cases. Please search for Stripette® pipettes when looking for smaller order quantities.