Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.8%, white crystalline powder for molecular biology
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:77-86-1
:
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
:121.14 g/mol
Frequently Bought Together