Order Entry
Czech Republic
ContactUsLinkComponent
Fig tree Ficin, MP Biomedicals
Fig tree Ficin, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Fig tree Ficin, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9001-33-6
  • Species:
    Fig tree
  • Storage conditions:
    −20 °C
  • Enzyme name:
    Ficin
  • Molecular weight:
    ~25 kDa (Lit.)
  • pH:
    5,0 - 7,5 (optimum pH for activity)

Specifications

Frequently Bought Together