Doxycycline hydrochloride
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:10592-13-9
:
Formula:C₂₂H₂₄N₂O₁₀·HCl
MDL Number:MFCD03427564
:480.9 g/mol
Frequently Bought Together
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Tubing, PTFE
Supplier: HAMILTON BONADUZ