Order Entry
Czech Republic
ContactUsLinkComponent
D-Glucose-6-phosphate disodium salt hydrate ≥98%
D-Glucose-6-phosphate disodium salt hydrate ≥98%
Supplier:  MP Biomedicals
Certificates
D-Glucose-6-phosphate disodium salt hydrate ≥98%
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity :
    ≥98%
  • FTIR:
    Conforms to Standard
  • Optical Rotation:
    +30° to +38° (c=1, water)
  • pH:
    7 - 9 (1% aq soln)

Specification Test Results

Frequently Bought Together