Order Entry
Czech Republic
ContactUsLinkComponent
 
Silicon dioxide, MP EcoChrom™, active
Catalog # ICNA0202769
Supplier:  MP Biomedicals
CAS Number:  
Silicon dioxide, MP EcoChrom™, active
Catalog # ICNA0202769
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :60.08 g/mol
Density:2.65 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232