ContactUsLinkComponent
General purpose gloves, Nitrotough™ N230B and N250B
General purpose gloves, Nitrotough™ N230B and N250B
Supplier:  Marigold (Ansell)
Certificates
General purpose gloves, Nitrotough™ N230B and N250B
Supplier:  Marigold (Ansell)
Specifications
  • Catalog No:
  • MARG200QB0580
  • MARG200QB0589
  • MARG200QB0588
  • Model:
  • N230B
  • N230B
  • N230B
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Cotton
  • Cotton
  • Cotton
  • Finish:
  • ¾ dipped
  • ¾ dipped
  • ¾ dipped
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4111A)
  • EN 388:2016 (4111A)
  • EN 388:2016 (4111A)