ContactUsLinkComponent
Labels for thermal transfer printers, printable, type B-7610
  BRDY800860
 :  Brady
undefined
Labels for thermal transfer printers, printable, type B-7610
  BRDY800860
 :  Brady
  • Type:
    B-7610
  • Colour:
    White
  • Description:
    Printer labels
  • Dimensions:
    80×50 mm
  • Material:
    Paper
  • Packed:
    750 labels/roll
  • Pk:
    4 Roll
  • Adhesive:
    Permanent acrylic
Paper thermal transfer printable labels for low cost general labelling applications.