ContactUsLinkComponent
Accessories
Accessories
Catalog # PALB96525700
Supplier:  Palbam
undefined
Accessories
Catalog # PALB96525700
Supplier:  Palbam
Specifications
  • Description:
    Telescopic arm for sensor carts, 3 m height extendable / 1 m horizontal extendable, made from stainless steel and anodized aluminum
  • Pk:
    1