Iron(II) gluconate hydrate ≥98%, powder, Sigma-Aldrich®
SIAL344427-25G
: Merck
:
- Pack type:Poly bottle
- Pk:25 g
- Seq:3308
- Appearance (Colour):White to Tan
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Iron (Fe)- Titration by Na2S2O3:≥11,3%
- Optical Rotation c = 1%; Water:3,7 - 9,7°
:
:464.16 g/mol
:188 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
:22830-45-1
:C₁₂H₂₄FeO₁₅
:MFCD00150872