Water, OmniPur® WFI cell culture tested, sterile, Calbiochem®
4.86505.9010
:
- Pack type:Plastic receptable in fibreboard box
- Pk:10 L
- Seq:5214
- Water conductivity (packaged water):Passes Test
- Water conductivity (prior to packaging, stage 1):Passes Test
- DNase:None detected
- Oxidisable substances:Passes Test
- Plating efficiency:Passes Test
- Protease:None detected
- RNase:None detected
- Particulate matter in injections:≤25 Particles/ml ≥10 µm in size
- Total Organic Carbon:500 ppb
- Sterility:Passes Test
- Bacterial endotoxin:≤0,025 EU/ml (per master product record)
:
:18.02 g/mol
:Ambient
:7732-18-5
:H₂O
:MFCD00011332