Order Entry
Czech Republic
ContactUsLinkComponent
Iodine monochloride solution 0.1 mol/l (0.2N) acc. Wijs for volumetric analysis
Iodine monochloride solution 0.1 mol/l (0.2N) acc. Wijs for volumetric analysis
Catalog # APPC281590.1611
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Iodine monochloride solution 0.1 mol/l (0.2N) acc. Wijs for volumetric analysis
Catalog # APPC281590.1611
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Seq:
    2865

Specifications

About this item

 :
 :162.36 g/mol
CAS Number:7790-99-0
Formula:ClI
MDL Number:MFCD00011354