MICROCHEM® 4000, 406 Chemical Resistant Boot Covers
Catalog # ANSEGR40W-00406-00
Supplier: Ansell
Specifications
- Shoe size:42 - 46
- Garment application:Chemical protection,Pharmaceutical
- Colour:Green
- Pk:100 PAIR
Specifications
About this item
These boot covers are made from a multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film) that is renowned for its lightweight, yet robust textile feel and exceptional barrier to organic & inorganic chemicals.
- Elastic to top of boot
- Tie fastening
- Ultrasonically welded seams
Typical applications include: Chemical handling, transportation, oil-based mud protection, HAZMAT emergency response, pharmaceutical, mining and agriculture.
Certifications: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5