ContactUsLinkComponent
ESD shoe covers, MICROGARD® 2000, model 417
ESD shoe covers, MICROGARD® 2000, model 417
Catalog # ANSEWH20B-00417-02
Supplier:  Ansell
undefined
ESD shoe covers, MICROGARD® 2000, model 417
Catalog # ANSEWH20B-00417-02
Supplier:  Ansell
Specifications
  • Shoe size:
    39 - 42
  • Garment application:
    Chemical protection
  • Colour:
    White
  • Seq:
    01
  • Pk:
    250 PAIR
Specifications
About this item
These over shoes are made from a microporous PE film with a non woven inner layer. This fabric provides low hazard liquid chemical repllence particle protection, protection from pesticides and a barrier to biological agents.
  • Elasticated opening
  • Bound seams
  • ESD PVC sole
  • Different sizes available