Cleaned foam cartridges
Supplier: Restek
Certificates
About this item
These precleaned cartridges are suitable for the air sampling during ambient, indoor and industrial hygiene applications.
- Ready to use for collection
- Made from cleaned PU foam
- PUF/XAD-2 sandwiches for semi-volatiles
Used in monitoring semi-volatiles organic compounds (SVOC) such as pesticides, PCB, and PAH.
Frequently Bought Together