


Cetrimonium bromide ≥98% for molecular biology
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:57-09-0
:
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772
:364.45 g/mol
Melting Pt:248…251 °C (lit.)
Flash Pt:244 °C (lit.)
EINECS:200-311-3
UN:3077
ADR:9,III
:GHS08
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Physical Appearance:Powder
- pH:6.0-7.5 (0.1 M H2O, 0 °C)(Lit.)
- Aggregation Number:170
- Critical Micelle Concentration:1 mM(20-25°C)
- Solubility:Freely but slowly soluble in water; Freely soluble in methanol and ethanol; soluble in alcohol.
Specification Test Results
Frequently Bought Together