Anti-CGA Mouse Monoclonal Antibody [clone: HCGab/52]
ABNOMAB14427
: Abnova
- Antibody type:Primary
- Antigen name:Native purified hCG protein
- Antigen symbol:CGA
- Clonality:Monoclonal
- Clone:HCGab/52
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:1081
- Storage buffer:In 10 mM PBS (0.05% BSA and 0.05% azide).
- Storage temperature:Store at 4° C.
- Immunogen:Native purified hCG protein.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against intact-HCG.
Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: HCGab/52
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human