Methyl-β-cyclodextrin (mixture of several Methylated)
:128446-36-6
:
:MFCD00074980
:1303.3000 g/mol
:180…182 °C
:Ambient
:411-120-1
- Appearance (Color):White to off-white
- Form:Powder
- Optical Rotation:+150,0 ° to +165,5 °, c-1,5 in water
- Solubility:Soluble in water
Frequently Bought Together