Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Bafilamycin A1
Catalog # J61835.MX
CAS Number:  
Irritant
Bafilamycin A1
Catalog # J61835.MX
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :622.83 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:88899-55-2
Formula:C₃₅H₅₈O₉
MDL Number:MFCD06795130