Anti-AFF4 Rabbit Polyclonal Antibody
Catalog # BSBTA03824
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:AFF4
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for AF4/FMR2 family member 4(AFF4) detection. Tested with WB in Human.
Type: Primary
Antigen: AFF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human