Environmental/HazMat cabinets, type 13/20
Dodavatel: CEMO
Certifikáty
O tomto produktu
These environmental/HazMat cabinet 13/20, based on the drum rack 540 serves for storage of environmentally harmful substances such as pesticides, lubricants, etc. The surface of cabinet casing is sealed with a high quality powder coating (RAL 7035 light grey).
- Closed with warning signs, doors with top and bottom ventilation slits
- With GRP sump 150 L, gratings and bottom tray
- Each level can support up to 200 kg
- Cabinet is delivered ready for assembly
These cabinet has a map case holder inside of the door in which packing notes, treatment instructions, etc can be properly stored. Also thr shelf depth of 600 mm enables horizontal storage of 60 L drums including discharge valve. The base frame and the grating floor are completely hot-dip galvanised. The Environmental/ HazMat cabinet 13/20 is optionally available with a 150 L GRP sump pallet or with a galvanised and welded tray bottom of 20 L capacity as the lowest storage level.
Frequently Bought Together