Order Entry
Czech Republic
ContactUsLinkComponent
Protexidase™, peroxidase protection buffer, Immuno™, MP Biomedicals
Protexidase™, peroxidase protection buffer, Immuno™, MP Biomedicals
Catalog # ICNA08980631
Supplier:  MP Biomedicals
CAS Number:  
Protexidase™, peroxidase protection buffer, Immuno™, MP Biomedicals
Catalog # ICNA08980631
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    Protexidase, peroxidase protection buffer
  • pH value:
    7,2

Specifications

About this item

A peroxidase protective buffer which stabilises HRP conjugates. It has 500 ml of PBS, pH 7,2, containing 0,5% BSA, antimicrobial preservatives and proprietary stabilising compounds.

It is compatible with all common blocking agents including Bovine Serum Albumin, skim milk powder and Polysorbate 20. Protexidase provides stability of the HRP enzyme and it also inhibits microbial growth in dilute conjugates.