Euro pallets and sumps, PE
Supplier: CEMO
Certificates
About this item
These sump pallets offer protection and a clean environment, through high resistance against acids and lyes and are used for the storage of caustic, flammable, explosive, firepromoting or toxic substances. Manufactured from high quality polyethylene (HDPE). For storing drums up to 224 L.
- High functionality and high chemical resistance
- Tray can be placed directly on the floor or on a Euro pallet
- Available in different versions with locking pins (for Euro-pallets), feet, runners or castors
Euro PE sump pallet (250/2 - RG) without approval no. are manufactured from high quality recycled polyethylene. Thet are suitable as a collection tray for materials not hazardous to water or for uses for which no approval is required.
Frequently Bought Together